Perfluorokerosene 50% in PFK low boiling, super-high boiling

Supplier: TCI America

Synonyms: PFK

P1061-5G
TCP1061-005GEA 268 USD
TCP1061-005G
Perfluorokerosene 50% in PFK low boiling, super-high boiling
Perfluorokerosene

[for Mass spectrometry]
MDL Number: MFCD00132835
Form: Clear Liquid
Color: Colorless
Flash Point (°C): 46


Product Categories: Analytical Science, Standards for Mass Spectrometry & Matrix Materials

https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=2b0c360d526c192334b2c9ac82377b767b22d4b8&vid=df839773bfaa5708f8b38962710b60e67ae427bd&prid=web&clid=SAPDAM

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR