Perfluorokerosene, super-high boiling

Supplier: TCI America

Synonyms: PFK

P1034-5G
TCP1034-005GEA 144 USD
TCP1034-005G
Perfluorokerosene, super-high boiling
Perfluorokerosene

[for Mass spectrometry]
MDL Number: MFCD00132835
Form: Clear Liquid
Color: Colorless
Specific Gravity (20/20): 2.05


Product Categories: Analytical Science, Standards for Mass Spectrometry & Matrix Materials

https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=a5b81698de5b70f9b0198295a8d11ba0e0af8128&vid=ab13b3efa9b648fa1b7c7a836386c54594c29f6c&prid=web&clid=SAPDAM

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR