Tris(2-ethylhexyl)phosphate ≥98.0%
Supplier: TCI America
CAS Number: 78-42-2
MDL Number: MFCD00009491
Molecular Formula: C24H51O4P
Molecular Weight: 434.64
Purity/Analysis Method:
Product Categories: Materials Chemistry, Secondary Battery Research Reagents, Resin Additives, Resin Additives
Other Product Names / Synonyms: Phosphoric Acid Trioctyl Ester, Phosphoric Acid Tris(2-ethylhexyl) Ester, Trioctyl Phosphate
Formula:
C₂₄H₅₁O₄P Boiling Pt: 215 °C Density: 0.93 (20 °C) Flash Pt: 204 °C |
MDL Number:
MFCD00009491 CAS Number: 78-42-2 EINECS: 201-116-6 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...