N-Phthaloylglycine ≥98.0%
Supplier: TCI America
Synonyms:
(1,3-Dihydro-1,3-dioxo-2H-isoindol-2-yl)acetic acid
P0963-500G
P0963-25G
TCP0963-500GEA
181
USD
TCP0963-500G
TCP0963-025G
N-Phthaloylglycine ≥98.0%
N-Phthaloylglycine
CAS Number: 4702-13-0
MDL Number: MFCD00005900
Molecular Formula: C10H7NO4
Molecular Weight: 205.17
Purity/Analysis Method:
Product Categories: Bioscience, Amino Acids (N-Protected)
Other Product Names / Synonyms: N-Phthalimidoacetic Acid
Formula:
C₁₀H₇NO₄ MW: 205.17 g/mol Melting Pt: 196 °C |
MDL Number:
MFCD00005900 CAS Number: 4702-13-0 |
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...