Pentafluoropyridine ≥98.0%
Supplier: TCI America
Synonyms:
2,3,4,5,6-Perfluoropyridine, 2,3,4,5,6-Pentafluoropyridine
P0926-25G
P0926-5G
TCP0926-25GEA
211
USD
TCP0926-25G
TCP0926-5G
Pentafluoropyridine ≥98.0%
Pentafluoropyridine
CAS Number: 700-16-3
MDL Number: MFCD00006225
Molecular Formula: C5F5N
Molecular Weight: 169.05
Purity/Analysis Method:
Product Categories: Synthetic Organic Chemistry, Fluorinated Building Blocks
Other Product Names / Synonyms: Perfluoropyridine
Formula:
C₅F₅N Boiling Pt: 83 °C Density: 1.62 (20 °C) Flash Pt: 25 °C |
MDL Number:
MFCD00006225 CAS Number: 700-16-3 UN: 1993 ADR: 3,III |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...