Pentafluorobenzene ≥98.0%

Supplier: TCI America

P0923-25G P0923-5G
TCP0923-25GEA 82 USD
TCP0923-25G TCP0923-5G
Pentafluorobenzene ≥98.0%
Pentafluorobenzene

CAS Number: 363-72-4 MDL Number: MFCD00000286 Molecular Formula: C6HF5 Molecular Weight: 168.07 Purity/Analysis Method: 98.0% (GC) Form: Clear Liquid Color: Colorless Boiling point (°C): 85 Melting point (°C): -48 Flash Point (°C): 13 Specific Gravity (20/20): 1.53

Formula: C₆HF₅
Boiling Pt: 85 °C
Melting Pt: –48 °C
Density: 1.53 (20 °C)
Flash Pt: 13 °C
MDL Number: MFCD00000286
CAS Number: 363-72-4
UN: 1993
ADR: 3,II
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=787ba3398d67be4f797749d6fde604a90cd5984d&vid=aca9ea41f8ffb3e5d010fa77264dddb73bf3f00b&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR