2-(Phenoxymethyl)benzoic acid ≥98.0%
Supplier: TCI America
P0905-25G
TCP0905-025GEA
146.5
USD
TCP0905-025G
2-(Phenoxymethyl)benzoic acid ≥98.0%
2-(Phenoxymethyl)benzoic acid
CAS Number: 724-98-1
MDL Number: MFCD00020294
Molecular Formula: C14H12O3
Molecular Weight: 228.25
Purity/Analysis Method:
Other Product Names / Synonyms: alpha-Phenoxy-o-toluic Acid
Formula:
C₁₄H₁₂O₃ Melting Pt: 126 °C |
MDL Number:
MFCD00020294 CAS Number: 724-98-1 EINECS: 211-967-5 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...