N,N,N',N',N''-Pentamethyldiethylenetriamine ≥99.0%

Supplier: TCI America

P0881-500ML P0881-25ML
TCP0881-500MLEA 106.53 USD
TCP0881-500ML TCP0881-25ML
N,N,N',N',N''-Pentamethyldiethylenetriamine ≥99.0%
N,N,N',N',N''-Pentamethyldiethylenetriamine

CAS Number: 3030-47-5 MDL Number: MFCD00014876 Molecular Formula: C9H23N3 Molecular Weight: 173.30 Purity/Analysis Method: 99.0% (GC) Form: Clear Liquid Color: Colorless Boiling point (°C): 86 Melting point (°C): -20 Flash Point (°C): 72 Specific Gravity (20/20): 0.83


Product Categories: Materials Chemistry, Precise Polymerization Reagents, Polymerization Catalysts Other Product Names / Synonyms: PMDTA, PMDETA

Formula: C₉H₂₃N₃
Boiling Pt: 86 °C
Melting Pt: –20 °C
Density: 0.83 (20 °C)
Flash Pt: 72 °C
MDL Number: MFCD00014876
CAS Number: 3030-47-5
EINECS: 221-201-1
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=032d66f35f2a1de03064e377294515ddeb952a24&vid=032d66f35f2a1de03064e377294515ddeb952a24&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 99.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR