1,10-Phenanthroline monohydrate ≥98.0% (by titrimetric analysis)

Supplier: TCI America

Synonyms: 1,10-Phenanthroline hydrate

P0879-25G
TCP0879-25GEA 98.5 USD
TCP0879-25G
1,10-Phenanthroline monohydrate ≥98.0% (by titrimetric analysis)
1,10-Phenanthroline monohydrate

CAS Number: 5144-89-8 MDL Number: MFCD00149973 Molecular Formula: C12H8N2 Molecular Weight: 180.21 Purity/Analysis Method: 98.0% (T) Form: Crystal Color: White Melting point (°C): 102

Formula: C₁₂H₈N₂·1H₂O
MW: 198.22 g/mol
Melting Pt: 102 °C
MDL Number: MFCD00011678
CAS Number: 5144-89-8
UN: 2811
ADR: 6.1,III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=160655fadcff37b08c57c69a14d6ba38368e9a1c&vid=160655fadcff37b08c57c69a14d6ba38368e9a1c&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (T)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR