Chloropentafluorobenzene ≥98.0%

Supplier: TCI America

Synonyms: Pentafluorochlorobenzene

P0850-25G
TCP0850-025GEA 70 USD
TCP0850-025G
Chloropentafluorobenzene ≥98.0%
Chloropentafluorobenzene

CAS Number: 344-07-0 MDL Number: MFCD00000534 Molecular Formula: C6ClF5 Molecular Weight: 202.51 Purity/Analysis Method: 98.0% (GC) Form: Clear Liquid Boiling point (°C): 117 Freezing point (°C): -15 Specific Gravity (20/20): 1.65

Formula: C₆ClF₅
Boiling Pt: 117 °C
Melting Pt: –15 °C
Density: 1.65 (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00000534
CAS Number: 344-07-0
EINECS: 206-450-6
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=4193229923b35827f5f1e7e2b7392874c1076f1a&vid=4193229923b35827f5f1e7e2b7392874c1076f1a&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR