Pivalaldehyde (trimethylacetaldehyde) ≥95.0%

Supplier: TCI America

Synonyms: Pivalaldehyde, Pivaldehyde, Trimethylacetaldehyde, 2,2-Dimethylpropionaldehyde, 2,2-Dimethylpropanal

P0847-25ML
TCP0847-025MLEA 190.91 USD
TCP0847-025ML
Pivalaldehyde (trimethylacetaldehyde) ≥95.0%
Pivalaldehyde (Trimethylacetaldehyde)

CAS Number: 630-19-3 MDL Number: MFCD00006962 Molecular Formula: C5H10O Molecular Weight: 86.13 Purity/Analysis Method: 95.0% (GC) Form: Clear Liquid Boiling point (°C): 75 Melting point (°C): 6 Flash Point (°C): 20 Specific Gravity (20/20): 0.78 Storage Temperature: 0-10°C


Product Categories: Synthetic Organic Chemistry, Aldehydes (Multi-component Reaction) Other Product Names / Synonyms: 2,2-Dimethylpropanal, 2,2-Dimethylpropionaldehyde, Trimethylacetaldehyde

Formula: C₅H₁₀O
Boiling Pt: 75 °C
Melting Pt: 6 °C
Density: 0.78 (20 °C)
Flash Pt: 20 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00006962
CAS Number: 630-19-3
EINECS: 211-134-6
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=f2a3258ca834f24655964dc5fd42c75f16224c81&vid=f2a3258ca834f24655964dc5fd42c75f16224c81&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 95.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR