Bis(2-ethylhexyl)phosphate ≥95.0%

Supplier: TCI America
Danger

Synonyms: Bis(2-ethylhexyl) hydrogen phosphate, Di-(2-ethylhexyl)phosphoric acid

P0811-500ML P0811-25ML
TCP0811-500MLEA 108.8 USD
TCP0811-500ML TCP0811-25ML
Bis(2-ethylhexyl)phosphate ≥95.0%
Bis(2-ethylhexyl) phosphate

[for Rare Metals Extraction] CAS Number: 298-07-7 MDL Number: MFCD00009492 Molecular Formula: C16H35O4P Molecular Weight: 322.43 Purity/Analysis Method: 95.0% (T) Form: Clear Liquid Melting point (°C): -50 Flash Point (°C): 196 Specific Gravity (20/20): 0.98


Product Categories: Materials Chemistry, Metal Extractants Other Product Names / Synonyms: Phosphoric Acid Di(2-ethylhexyl) Ester, Di(2-ethylhexyl) Phosphate, Dioctyl Phosphate, Phosphoric Acid Dioctyl Ester

Formula: C₁₆H₃₅O₄P
Melting Pt: –50 °C
Density: 0.98 (20 °C)
Flash Pt: 196 °C
MDL Number: MFCD00009492
CAS Number: 298-07-7
EINECS: 206-056-4
UN: 1902
ADR: 8,III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=2d72f139b09afa5f0f3ce2e0a74387fe998e106c&vid=0c5ac6429daab9153bdb02e6bdbd69fdd87d2d5f&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 95.0% (T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR