(S)-(-)-α-Methylbenzylamine ≥98.0%

Supplier: TCI America
Danger

P0793-500ML P0793-25ML P0793-100ML
TCP0793-500MLEA 372.31 USD
TCP0793-500ML TCP0793-25ML TCP0793-100ML
(S)-(-)-α-Methylbenzylamine ≥98.0%
(S)-(-)-α-Methylbenzylamine

CAS Number: 2627-86-3 MDL Number: MFCD00064406 Molecular Formula: C8H11N Molecular Weight: 121.18 Purity/Analysis Method: 98.0% (GC) Form: Clear Liquid Boiling point (°C): 84 Flash Point (°C): 79 Specific Gravity (20/20): 0.95 Specific rotation [a]20/D: -39 deg (neat)


Product Categories: Synthetic Organic Chemistry, Optical Resolving Agents, Chiral Building Blocks, Analytical Science, e.e. / Absolute Configuration Determination (NMR) Other Product Names / Synonyms: (S)-(-)-alpha-Methylbenzylamine

Formula: C₈H₁₁N
Boiling Pt: 84 °C
Melting Pt: –10 °C
Density: 0.95 (20 °C)
Flash Pt: 79 °C
Storage Temperature: Ambient
MDL Number: MFCD00064406
CAS Number: 2627-86-3
EINECS: 220-098-0
Merck Index: 12,06106
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=38ce516b11b5e2fa12b9ee5378ea97b92c26d51e&vid=38ce516b11b5e2fa12b9ee5378ea97b92c26d51e&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR