Polyethylene glycol monostearate n=∼10

Supplier: TCI America

P0719-500G P0719-25G
TCP0719-500GEA 86 USD
TCP0719-500G TCP0719-025G
Polyethylene glycol monostearate n=∼10
Polyethylene glycol monostearate

Palmitate and stearate mixture


Form: Crystal
Color: White

Formula: HO(CH₂CH₂O)nOCC₁₇H₃₅
Flash Pt: 266 °C
MDL Number: MFCD00148007
CAS Number: 9004-99-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR