Polyethylene glycol monostearate n=∼4

Supplier: TCI America

P0718-25G P0718-500G
TCP0718-025GEA 21.37 USD
TCP0718-025G TCP0718-500G
Polyethylene glycol monostearate n=∼4
Polyethylene glycol monostearate

Palmitate and stearate mixture


Form: Crystal
Color: White

Formula: HO(CH₂CH₂O)nOCC₁₇H₃₅
MDL Number: MFCD00148007
CAS Number: 9004-99-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR