Phenolphthalein 0,1% in ethanol 85%

Supplier: TCI America
Danger

Synonyms: 3,3-Bis(4-hydroxyphenyl)isobenzofuran-1(3H)-one

P0700-100ML P0700-500ML
TCP0700-100MLEA 23.06 USD
TCP0700-100ML TCP0700-500ML
Phenolphthalein 0,1% in ethanol 85%
Phenolphthalein

[for Titration]
CAS Number: 77-09-8
MDL Number: MFCD00005913
Molecular Weight: 318.33
Form: Clear Liquid
Color: Colorless
Flash Point (°C): 13

Formula: C₂₀H₁₄O₄
Boiling Pt: ∼78 °C (1013 hPa)
Density: ∼0.85 g/cm³ (20 °C)
Flash Pt: 13 °C
Storage Temperature: Ambient
MDL Number: MFCD00005913
CAS Number: 77-09-8
EINECS: 201-004-7
ADR: 3,II
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=f46b1a7aa2d7577ea9ebe10c72b98bc1795515cb&vid=d66f2ff872973a70d207f8d6cc92ca3595da125e&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR