Perfluorokerosene, low boiling

Supplier: TCI America

Synonyms: PFK

P0690-10G
TCP0690-010GEA 180 USD
TCP0690-010G
Perfluorokerosene, low boiling
Perfluorokerosene

[for Mass spectrometry]
MDL Number: MFCD00132835
Form: Clear Liquid
Color: Colorless
Flash Point (°C): 46
Specific Gravity (20/20): 1.86


Product Categories: Analytical Science, Standards for Mass Spectrometry & Matrix Materials

https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=116a12ead0ed56c97bbd63234ef1670cdacb369d&vid=ff76e9f219e6c63a94ceb39d9149d15c1dbaf358&prid=web&clid=SAPDAM

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR