Lithium pyruvate monohydrate ≥95.0% (by titrimetric analysis)
Supplier: TCI America
[Guaranteed for Standard to GOT, GPT]
CAS Number: 2922-61-4
MDL Number: MFCD00036136
Molecular Formula: C3H4O3
Molecular Weight: 93.99
Purity/Analysis Method:
Product Categories: Materials Chemistry, Secondary Battery Research Reagents
Other Product Names / Synonyms: Pyruvic Acid Lithium Salt
Formula:
C₃H₄O₃ |
MDL Number:
MFCD00036136 CAS Number: 2922-61-4 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...