4-Pentenoic acid ≥98.0% (by GC, titration analysis)

Supplier: TCI America

P0645-25ML
TCP0645-25MLEA 80 USD
TCP0645-25ML
4-Pentenoic acid ≥98.0% (by GC, titration analysis)
4-Pentenoic acid

CAS Number: 591-80-0 MDL Number: MFCD00004408 Molecular Formula: C5H8O2 Molecular Weight: 100.12 Purity/Analysis Method: 98.0% (GC,T) Form: Clear Liquid Boiling point (°C): 182 Flash Point (°C): 93 Specific Gravity (20/20): 0.98


Other Product Names / Synonyms: Allylacetic Acid, 3-Vinylpropionic Acid

Formula: C₅H₈O₂
Boiling Pt: 182 °C
Density: 0.98 (20 °C)
Flash Pt: 93 °C
MDL Number: MFCD00004408
CAS Number: 591-80-0
UN: 3265
ADR: 8,III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=cac82daf62f92cc6ac982ba5c959e9a499a87c0a&vid=cac82daf62f92cc6ac982ba5c959e9a499a87c0a&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC,T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR