4-Phenylbutyric acid ≥98.0%

Supplier: TCI America

Synonyms: 4-Phenylbutanoic acid

P0643-250G P0643-25G
TCP0643-250GEA 310.7 USD
TCP0643-250G TCP0643-025G
4-Phenylbutyric acid ≥98.0%
4-Phenylbutyric acid

CAS Number: 1821-12-1 MDL Number: MFCD00004403 Molecular Formula: C10H12O2 Molecular Weight: 164.20 Purity/Analysis Method: 98.0% (GC,T) Form: Crystal Boiling point (°C): 175 Melting point (°C): 52 Flash Point (°C): 100

Formula: C₁₀H₁₂O₂
Boiling Pt: 175 °C
Melting Pt: 52 °C
Flash Pt: 100 °C
MDL Number: MFCD00004403
CAS Number: 1821-12-1
EINECS: 217-341-8
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=d593e3c252db6e92e8166d558ca31ae679f03a15&vid=d593e3c252db6e92e8166d558ca31ae679f03a15&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC,T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR