Purpurin(1,2,4-trihydroxy-9,10-anthraquinone) ≥95.0% (by HPLC)
Supplier: TCI America
CAS Number: 81-54-9
MDL Number: MFCD00001203
Molecular Formula: C14H8O5
Molecular Weight: 256.21
Purity/Analysis Method:
Product Categories: Bioscience, Others (Phytochemicals)
Other Product Names / Synonyms: 1,2,4-Trihydroxyanthraquinone
Formula:
C₁₄H₈O₅ Melting Pt: 257 °C |
MDL Number:
MFCD00001203 CAS Number: 81-54-9 EINECS: 201-359-8 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...