Sudan I ≥95.0% (by HPLC)

Supplier: TCI America

Synonyms: Scarlet B, 1-(Phenyldiazenyl)naphthalen-2-ol, 1-Phenylazo-2-naphthol, Oil Orange, Solvent Yellow 14, Sudan Yellow

P0585-25G
TCP0585-025GEA 27 USD
TCP0585-025G
Sudan I ≥95.0% (by HPLC)
Sudan I

CAS Number: 842-07-9 MDL Number: MFCD00003911 Molecular Formula: C16H12N2O Molecular Weight: 248.29 Purity/Analysis Method: 95.0% (HPLC) Form: Crystal Melting point (°C): 129 Lambda max.: 480 nm (MeOH)


Product Categories: Bioscience, Stains & Dyes Other Product Names / Synonyms: Oil Orange, 1-Phenylazo-2-naphthol, Scarlet B, Solvent Yellow 14, Sudan Yellow

Formula: C₁₆H₁₂N₂O
MW: 248.28 g/mol
Boiling Pt: 444 °C (1013 hPa)
Melting Pt: 129 °C
Density: 1.119 g/cm³ (151 °C)
MDL Number: MFCD00003911
CAS Number: 842-07-9
EINECS: 212-668-2
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=ffc620d9b285fb24484c4221c4e70a20d8432f5b&vid=ffc620d9b285fb24484c4221c4e70a20d8432f5b&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 95.0% (HPLC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR