2-Pyrrolidone ≥98.0%

Supplier: TCI America

P0575-25G P0575-500G
TCP0575-025GEA 21.08 USD
TCP0575-025G TCP0575-500G
2-Pyrrolidone ≥98.0%
2-Pyrrolidone

CAS Number: 616-45-5 MDL Number: MFCD00005270 Molecular Formula: C4H7NO Molecular Weight: 85.11 Purity/Analysis Method: 98.0% (GC) Form: Crystal Boiling point (°C): 245 Flash Point (°C): 145 Freezing point (°C): 24


Other Product Names / Synonyms: 2-Azacyclopentanone, gamma-Butyrolactam, 2-Ketopyrrolidine, 2-Pyrrolidinone

Formula: C₄H₇NO
Boiling Pt: 245 °C
Melting Pt: 24…26 °C
Density: 1.1115 g/cm³ (20 °C)
Flash Pt: 145 °C
Storage Temperature: Ambient
MDL Number: MFCD00005270
CAS Number: 616-45-5
EINECS: 210-483-1
Merck Index: 12,08200
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=40381f263d2ec9a2897e3018f3883cf9507a343c&vid=40381f263d2ec9a2897e3018f3883cf9507a343c&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR