Pyrrole ≥99.0%

Supplier: TCI America
Danger

Synonyms: 1H-Pyrrole

P0574-500ML P0574-25ML P0574-100ML
TCP0574-500MLEA 319.3 USD
TCP0574-500ML TCP0574-25ML TCP0574-100ML
Pyrrole ≥99.0%
Pyrrole

CAS Number: 109-97-7 MDL Number: MFCD00005216 Molecular Formula: C4H5N Molecular Weight: 67.09 Purity/Analysis Method: 99.0% (GC) Form: Clear Liquid Boiling point (°C): 130 Melting point (°C): -23 Flash Point (°C): 39 Specific Gravity (20/20): 0.97


Product Categories: Materials Chemistry, Conducting Polymer Research Reagents Other Product Names / Synonyms: Divinylenimine

Formula: C₄H₅N
MW: 67.09 g/mol
Boiling Pt: 130 °C
Melting Pt: -24
Density: 0.97 (20 °C)
Flash Pt: 39
MDL Number: MFCD00005216
CAS Number: 109-97-7
EINECS: 203-724-7
UN: 1992
ADR: 3 III
Merck Index: 12,08198
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=a7b298b28560533d1ffd65adb1866e19585e2b3c&vid=a7b298b28560533d1ffd65adb1866e19585e2b3c&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 99.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR