(±)-1,2-Propanediol ≥99.0% (by GC)

Supplier: TCI America
Warning

Synonyms: Propylene glycol, 1,2-Propanediol, (±)-1,2-Propylene glycol, 1,2-Dihydroxypropane, (±)-1,2-Dihydroxy propane

P0485-500G
TCP0485-500GEA 34 USD
TCP0485-500G
(±)-1,2-Propanediol ≥99.0% (by GC)
Propylene glycol
Formula: CH₃CH(OH)CH₂OH
MW: 76.1 g/mol
Boiling Pt: 188°C
Melting Pt: -59°C
Flash Pt: 107°C
MDL Number: MFCD00064272
CAS Number: 57-55-6
EINECS: 200-338-0
Merck Index: 14,07855

Order Now

Specification Test Results

Appearance Colorless to Almost colorless clear liquid
Purity (GC) min. 99.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR