Polyvinylpyrrolidone, M.W. K 15 (MW ∼ 10,000)
Supplier: TCI America
P0471-25G
P0471-500G
P0471-100G
TCP0471-025GEA
32.33
USD
TCP0471-025G
TCP0471-500G
TCP0471-100G
Polyvinylpyrrolidone, M.W. K 15 (MW ∼ 10,000)
Polyvinylpyrrolidone
CAS Number: 9003-39-8
Form: Powder
Color: White
Flash Point (°C): 215
Product Categories: Materials Chemistry, Polymer Research Reagents
Formula:
(C₆H₉NO)n Storage Temperature: Ambient |
MDL Number:
MFCD00149016 CAS Number: 9003-39-8 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...