Pentaethylene glycol monododecyl ether ≥98.0%
Supplier: TCI America
P0372-5G
TCP0372-5GEA
1732
USD
TCP0372-5G
Pentaethylene glycol monododecyl ether ≥98.0%
Pentaethylene glycol monododecyl ether
CAS Number: 3055-95-6
MDL Number: MFCD00043064
Molecular Formula: C22H46O6
Molecular Weight: 406.60
Purity/Analysis Method:
Formula:
C₂₂H₄₆O₆ |
MDL Number:
MFCD00043064 CAS Number: 3055-95-6 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...