Phthalocyanine ≥93.0% (by total nitrogen basis)

Supplier: TCI America

Synonyms: Pigment Blue 16, 29H,31H-Phthalocyanine

P0355-25G
TCP0355-025GEA 109.8 USD
TCP0355-025G
Phthalocyanine ≥93.0% (by total nitrogen basis)
Phthalocyanine

CAS Number: 574-93-6 MDL Number: MFCD00005085 Molecular Formula: C32H18N8 Molecular Weight: 514.55 Purity/Analysis Method: 93.0% (N) Form: Crystal


Product Categories: Synthetic Organic Chemistry, Ligands, Materials Chemistry, Molecular Conductors, Porphyrin & Phthalocyanine Dyes (Functional Organic Dyes), Building Blocks for Phthalocyanines Other Product Names / Synonyms: Pigment Blue 16

Formula: C₃₂H₁₈N₈
MW: 514.55 g/mol
Melting Pt: 210 °C
Density: 1.447 g/cm³ (–113 °C)
MDL Number: MFCD00005085
CAS Number: 574-93-6
EINECS: 209-378-3
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=2c3466dfc7ae40a3b3c9fdca0108f2e093d6c93a&vid=2c3466dfc7ae40a3b3c9fdca0108f2e093d6c93a&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 93.0% (N)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR