Tritolyl phosphate (mixture of isomers) ≥99.0% (by GC)

Supplier: TCI America
Warning

P0273-500G P0273-25G
TCP0273-500GEA 45 USD
TCP0273-500G TCP0273-25G
Tritolyl phosphate (mixture of isomers) ≥99.0% (by GC)
Tritolyl phosphate (mixture of isomeres)

CAS Number: 1330-78-5 MDL Number: MFCD02102116 Molecular Formula: C21H21O4P Molecular Weight: 368.37 Purity/Analysis Method: 99.0% (GC) Form: Clear Liquid Boiling point (°C): 255 Melting point (°C): -33 Flash Point (°C): 234 Specific Gravity (20/20): 1.17


Product Categories: Materials Chemistry, Secondary Battery Research Reagents, Resin Additives, Resin Additives Other Product Names / Synonyms: Phosphoric Acid Tritolyl Ester, Phosphoric Acid Tricresyl Ester, Tritolyl Phosphate

Formula: C₂₁H₂₁O₄P
Boiling Pt: 255 °C
Melting Pt: –33 °C
Density: 1.17 (20 °C)
Flash Pt: 234 °C
MDL Number: MFCD02102116
CAS Number: 1330-78-5
EINECS: 215-548-8
UN: 2810
ADR: 6.1,II
Merck Index: 12,09892
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=4443dfbb4c5ddb084f9c39f864f5080bc124c47d&vid=4443dfbb4c5ddb084f9c39f864f5080bc124c47d&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Appearance Colorless to Light yellow clear liquid
Purity (GC) min. 99.0% (total of isomer)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR