L-Phenylalanine ≥98.0% (by HPLC, titration analysis)
Supplier: TCI America
CAS Number: 63-91-2
MDL Number: MFCD00064227
Molecular Formula: C9H11NO2
Molecular Weight: 165.19
Purity/Analysis Method:
Product Categories: Synthetic Organic Chemistry, Asymmetric Organocatalysts, Bioscience, Proteinogenic Amino Acids
Other Product Names / Synonyms: H-Phe-OH
Formula:
C₉H₁₁NO₂ Boiling Pt: 313 °C (1013 hPa) Melting Pt: 275 °C Density: 0.754 g/cm³ (20 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00064227 CAS Number: 63-91-2 EINECS: 200-568-1 Merck Index: 14,07271 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...