Phenol red

Supplier: TCI America

Synonyms: 3,3-Bis(4-hydroxyphenyl)-3H-benzo[c][1,2]oxathiole 1,1-dioxide

P0100-1G P0100-25G P0100-100G
TCP0100-001GEA 19.03 USD
TCP0100-001G TCP0100-025G TCP0100-100G
Phenol red
Phenol red

CAS Number: 143-74-8 MDL Number: MFCD00003552 Molecular Formula: C19H14O5S Molecular Weight: 354.38 Form: Crystal Color: Deep Yellow Red


Other Product Names / Synonyms: Phenolsulfonphthalein

Formula: C₁₉H₁₄O₅S
Melting Pt: > 300 °C
Storage Temperature: Ambient
MDL Number: MFCD00003552
CAS Number: 143-74-8
EINECS: 205-609-7
Merck Index: 13,07329
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=6e51da0f259217a65cf4d8881c4c62bba3ec1269&vid=6e51da0f259217a65cf4d8881c4c62bba3ec1269&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR