Phenolphthalein ≥98.0%

Supplier: TCI America
Danger

Synonyms: 3,3-Bis(4-hydroxyphenyl)isobenzofuran-1(3H)-one

P0094-25G P0094-500G
TCP0094-025GEA 28.23 USD
TCP0094-025G TCP0094-500G
Phenolphthalein ≥98.0%
Phenolphthalein

CAS Number: 77-09-8 MDL Number: MFCD00005913 Molecular Formula: C20H14O4 Molecular Weight: 318.33 Purity/Analysis Method: 98.0% (T) Form: Crystalline Powder Melting point (°C): 262

Formula: C₂₀H₁₄O₄
Melting Pt: 262 °C
Density: 1.3 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00005913
CAS Number: 77-09-8
EINECS: 201-004-7
REACH: 01-2119498295-24
Merck Index: 14,07243
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=5fdca9fd7fb790d5fed2f4aa5c40bef267f64fbb&vid=5fdca9fd7fb790d5fed2f4aa5c40bef267f64fbb&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR