2-Pentanone ≥99.0%
Supplier: TCI America
|
Danger
|
Synonyms:
Methyl Propyl Ketone
P0060-500ML
P0060-25ML
TCP0060-500MLEA
117.76
USD
TCP0060-500ML
TCP0060-25ML
2-Pentanone ≥99.0%
2-Pentanone
CAS Number: 107-87-9
MDL Number: MFCD00009400
Molecular Formula: C5H10O
Molecular Weight: 86.13
Purity/Analysis Method:
Other Product Names / Synonyms: Methyl Propyl Ketone
Formula:
C₅H₁₀O Boiling Pt: 101 °C Melting Pt: –78 °C Density: 0.81 (20 °C) Flash Pt: 7 °C Storage Temperature: Ambient |
MDL Number:
MFCD00009400 CAS Number: 107-87-9 EINECS: 203-528-1 UN: 1249 ADR: 3,II Merck Index: 12,06193 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...