1-Pentanol ≥99.0%

Supplier: TCI America
Danger

Synonyms: 1-Pentyl alcohol, 1-Hydroxy pentane, n-Amyl alcohol, Amyl alcohol, Butyl carbinol

P0055-500ML P0055-25ML
TCP0055-500MLEA 42.59 USD
TCP0055-500ML TCP0055-25ML
1-Pentanol ≥99.0%
1-Pentanol

CAS Number: 71-41-0 MDL Number: MFCD00002977 Molecular Formula: C5H12O Molecular Weight: 88.15 Purity/Analysis Method: 99.0% (GC) Form: Clear Liquid Boiling point (°C): 137 Melting point (°C): -79 Flash Point (°C): 43 Specific Gravity (20/20): 0.82


Product Categories: Materials Chemistry, Derivatizing Reagents for Solubility Improvement, Analytical Science, Odor Deterioration Factors of Cosmetics Other Product Names / Synonyms: Amyl Alcohol, Butylcarbinol

Formula: H₃C(CH₂)₃CH₂OH
Boiling Pt: 137 °C
Melting Pt: –79 °C
Density: 0.82 (20 °C)
Flash Pt: 43 °C
Storage Temperature: Ambient
MDL Number: MFCD00002977
CAS Number: 71-41-0
EINECS: 200-752-1
UN: 1105
ADR: 3,III
Merck Index: 13,07195
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=14bb2c139339e85f43ccf9c77bced226ece2caaa&vid=a4bfbda0c57603519c4d829b37947a303c3ae6a2&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 99.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR