Quercetin hydrate ≥95.0% (by HPLC and UV method)
Supplier: TCI America
Synonyms:
3,3',4',5,7-Pentahydroxyflavone hydrate, 2-(3,4-Dihydroxyphenyl)-3,5,7-trihydroxy-4H-chromen-4-one xhydrate
P0042-25G
TCP0042-025GEA
57.5
USD
TCP0042-025G
Quercetin hydrate ≥95.0% (by HPLC and UV method)
Quercetin hydrate
CAS Number: 849061-97-8
MDL Number: MFCD00006828
Molecular Formula: C15H10O7
Molecular Weight: 302.24
Purity/Analysis Method:
Product Categories: Bioscience, Aromatic Polyketides, Polyphenols (Phytochemicals), Histone Deacetylase Activators
Other Product Names / Synonyms: 3,3',4',5,7-Pentahydroxyflavone
Formula:
C₁₅H₁₀O₇ MW: 302.24 g/mol (anhy) |
MDL Number:
MFCD00006828 CAS Number: 849061-97-8 |
Specification Test Results
Purity/Analysis Method |
|
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...