Calcium pantothenate ≥98.0%

Supplier: TCI America

Synonyms: Calcium D-Pantothenate, D-Pantothenic acid calcium salt, D-Calcium pantothenate, Calcium (R)-3-(2,4-dihydroxy-3,3-dimethylbutanamido)propanoate, Vitamin B5 calcium salt, Calcium D-(+)-pantothenate

P0012-25G P0012-500G P0012-100G
TCP0012-025GEA 38.84 USD
TCP0012-025G TCP0012-500G TCP0012-100G
Calcium pantothenate ≥98.0%
Calcium pantothenate

CAS Number: 137-08-6 MDL Number: MFCD00002766 Molecular Formula: C9H17NO5 Molecular Weight: 476.54 Purity/Analysis Method: 98.0% (T) Form: Crystal Melting point (°C): 172 Specific rotation [a]20/D: 27 deg (C=5, H2O)


Product Categories: Bioscience, Vitamins Other Product Names / Synonyms: D-Pantothenic Acid Calcium Salt

Formula: C₁₈H₃₂CaN₂O₁₀
MW: 476.54 g/mol
Melting Pt: 172 °C
MDL Number: MFCD00002766
CAS Number: 137-08-6
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=ead89bbd860a59a50d9b47a4e136ddead5ba6eab&vid=ead89bbd860a59a50d9b47a4e136ddead5ba6eab&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR