Sodium thiosulfate pentahydrate ≥99.0% (by titrimetric analysis)

Supplier: TCI America

O0522-300G
TCO0522-300GEA 21 USD
TCO0522-300G
Sodium thiosulfate pentahydrate ≥99.0% (by titrimetric analysis)
Sodium thiosulphate pentahydrate
Formula: Na₂S₂O₃.5H₂O
MW: 248.19 g/mol
Melting Pt: 48 °C
MDL Number: MFCD00149186
CAS Number: 10102-17-7

Order Now

Specification Test Results

Appearance White powder to crystal
Purity (Iodometric Titration) min. 99.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR