Oxyresveratrol ≥96.0% (by GC)
Supplier: TCI America
CAS Number: 29700-22-9
MDL Number: MFCD11974969
Molecular Formula: C14H12O4
Molecular Weight: 244.25
Purity/Analysis Method:
Product Categories: Bioscience, Inhibitors (for Pharmacological Research), Antioxidants, Aromatic Polyketides, Polyphenols (Phytochemicals)
Other Product Names / Synonyms: 2,3',4,5'-Tetrahydroxy-trans-stilbene
Formula:
C₁₄H₁₂O₄ Melting Pt: 197 °C |
MDL Number:
MFCD11974969 CAS Number: 29700-22-9 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...