Octafluoroadipic acid ≥98.0%

Supplier: TCI America

Synonyms: Perfluoroadipic Acid, Octafluorohexanedioic Acid, Perfluorohexanedioic Acid

O0260-5G O0260-25G
TCO0260-005GEA 52 USD
TCO0260-005G TCO0260-25G
Octafluoroadipic acid ≥98.0%
Octafluoroadipic acid

CAS Number: 336-08-3 MDL Number: MFCD00066405 Molecular Formula: C6H2F8O4 Molecular Weight: 290.07 Purity/Analysis Method: 98.0% (GC,T) Form: Crystal Melting point (°C): 134


Product Categories: Synthetic Organic Chemistry, Fluorous Compounds, Fluorinated Building Blocks Other Product Names / Synonyms: Octafluorohexanedioic Acid, Perfluoroadipic Acid, Perfluorohexanedioic Acid

Formula: C₆H₂F₈O₄
Melting Pt: 134 °C
MDL Number: MFCD00066405
CAS Number: 336-08-3
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=29b6ee80a5c7cd85f0de9a0fd96322ff444544e4&vid=29b6ee80a5c7cd85f0de9a0fd96322ff444544e4&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC,T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR