2-Oxazolidinone ≥98.0%

Supplier: TCI America

Synonyms: 2-Oxazolidone, Oxazolidin-2-one

O0188-500G O0188-25G O0188-100G
TCO0188-500GEA 377.16 USD
TCO0188-500G TCO0188-025G TCO0188-100G
2-Oxazolidinone ≥98.0%
2-Oxazolidinone

CAS Number: 497-25-6 MDL Number: MFCD00005268 Molecular Formula: C3H5NO2 Molecular Weight: 87.08 Purity/Analysis Method: 98.0% (GC) Form: Crystal Boiling point (°C): 200 Melting point (°C): 89

Formula: C₃H₅NO₂
MW: 87.08 g/mol
Boiling Pt: 200 °C
Melting Pt: 89 °C
MDL Number: MFCD00005268
CAS Number: 497-25-6
EINECS: 207-840-9
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=3a812a1ad52998270b972dfb4d8e451dc90384bf&vid=3a812a1ad52998270b972dfb4d8e451dc90384bf&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR