(±)-1-Octen-3-ol ≥98.0%

Supplier: TCI America

Synonyms: (±)-Oct-1-en-3-ol, (±)-Pentyl vinyl carbinol, Oct-1-en-3-ol

O0159-25ML O0159-100ML
TCO0159-25MLEA 45.89 USD
TCO0159-25ML TCO0159-100ML
(±)-1-Octen-3-ol ≥98.0%
(±)-1-Octen-3-ol

CAS Number: 3391-86-4 MDL Number: MFCD00004589 Molecular Formula: C8H16O Molecular Weight: 128.22 Purity/Analysis Method: 98.0% (GC) Form: Clear Liquid Boiling point (°C): 166 Flash Point (°C): 63 Specific Gravity (20/20): 0.84

Formula: C₈H₁₆O
MW: 128.21 g/mol
Boiling Pt: 174
Melting Pt: -49
Density: 0.84 (20 °C)
Flash Pt: 63 °C
MDL Number: MFCD00004589
CAS Number: 3391-86-4
UN: 2810
ADR: 6.1 III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=bf21efc73d03cd1c823e1fd8ab401b19c8a7363c&vid=bf21efc73d03cd1c823e1fd8ab401b19c8a7363c&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR