cis-4-Octene ≥95.0%

Supplier: TCI America

Synonyms: (Z)-4-Octene

O0135-5ML
TCO0135-005MLEA 141 USD
TCO0135-005ML
cis-4-Octene ≥95.0%
cis-4-Octene

CAS Number: 7642-15-1 MDL Number: MFCD00067484 Molecular Formula: C8H16 Molecular Weight: 112.22 Purity/Analysis Method: 95.0% (GC) Form: Clear Liquid Boiling point (°C): 122 Flash Point (°C): 17 Specific Gravity (20/20): 0.72

Formula: C₈H₁₆
MW: 112.22 g/mol
Boiling Pt: 123
Melting Pt: -118.7
Density: 0.72 (20 °C)
Flash Pt: 17 °C
MDL Number: MFCD00067484
CAS Number: 7642-15-1
EINECS: 231-578-4
UN: 3295
ADR: 3 II

Order Now

Specification Test Results

Purity/Analysis Method 95.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR