4-Octyne ≥99.0%

Supplier: TCI America

O0127-5ML
TCO0127-005MLEA 44 USD
TCO0127-005ML
4-Octyne ≥99.0%
4-Octyne

CAS Number: 1942-45-6 MDL Number: MFCD00009471 Molecular Formula: C8H14 Molecular Weight: 110.20 Purity/Analysis Method: 99.0% (GC) Form: Clear Liquid Boiling point (°C): 131 Melting point (°C): -103 Flash Point (°C): 29 Specific Gravity (20/20): 0.75

Formula: C₈H₁₄
Boiling Pt: 131 °C
Melting Pt: –103 °C
Density: 0.75 (20 °C)
Flash Pt: 29 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00009471
CAS Number: 1942-45-6
EINECS: 217-730-2
UN: 1993
ADR: 3,II

Order Now

Specification Test Results

Purity/Analysis Method 99.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR