3-Octanone ≥98.0%

Supplier: TCI America

O0122-500ML O0122-25ML
TCO0122-500MLEA 185.13 USD
TCO0122-500ML TCO0122-25ML
3-Octanone ≥98.0%
3-Octanone

CAS Number: 106-68-3 MDL Number: MFCD00009515 Molecular Formula: C8H16O Molecular Weight: 128.22 Purity/Analysis Method: 98.0% (GC) Form: Clear Liquid Boiling point (°C): 167 Flash Point (°C): 54 Specific Gravity (20/20): 0.82


Product Categories: Analytical Science, Odor Deterioration Factors of Cosmetics Other Product Names / Synonyms: Amyl Ethyl Ketone

Formula: C₈H₁₆O
Boiling Pt: 167 °C
Melting Pt: –18.5 °C
Density: 0.82 (20 °C)
Flash Pt: 54 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00009515
CAS Number: 106-68-3
EINECS: 203-423-0
UN: 1224
ADR: 3,III
Merck Index: 13,03799
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=ca2caa90d4e1dcf136bd17ff7cdb6eef1d912e20&vid=81cdb3d50234e3792da381717e7be822d17f6769&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR