Diethyl oxalate ≥99.0%

Supplier: TCI America
Warning

Synonyms: Oxalic acid diethyl ester

O0120-250ML
TCO0120-250MLEA 119 USD
TCO0120-250ML
Diethyl oxalate ≥99.0%
Diethyl oxalate

[for Spectrophotometry] CAS Number: 95-92-1 MDL Number: MFCD00009119 Molecular Formula: C6H10O4 Molecular Weight: 146.14 Purity/Analysis Method: 99.0% (GC) Form: Clear Liquid Boiling point (°C): 185 Melting point (°C): -41 Flash Point (°C): 76 Specific Gravity (20/20): 1.08


Product Categories: Analytical Science, Solvents for Spectrophotometry Other Product Names / Synonyms: Oxalic Acid Diethyl Ester

Formula: C₆H₁₀O₄
MW: 146.14 g/mol
Boiling Pt: 185 °C
Melting Pt: –41 °C
Density: 1.08 (20 °C)
Flash Pt: 76 °C
Storage Temperature: Ambient
MDL Number: MFCD00009119
CAS Number: 95-92-1
EINECS: 202-464-1
UN: 2525
ADR: 6.1,III
Merck Index: 13,03152
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=8d375962fe29e33bdfc9d417b24c235135349ee6&vid=b434343e3bf3a706577d6a3c8f9323ad17e7f641&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 99.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR