n-Octane ≥98.5%

Supplier: TCI America
Danger

O0118-100ML
TCO0118-100MLEA 302.19 USD
TCO0118-100ML
n-Octane ≥98.5%
n-Octane

[for Spectrophotometry] CAS Number: 111-65-9 MDL Number: MFCD00009556 Molecular Formula: C8H18 Molecular Weight: 114.23 Purity/Analysis Method: 98.5% (GC) Form: Clear Liquid Boiling point (°C): 125 Melting point (°C): -57 Flash Point (°C): 14 Specific Gravity (20/20): 0.70

Formula: H₃C(CH₂)₆CH₃
MW: 114.23 g/mol
Boiling Pt: 125 °C
Melting Pt: –57 °C
Density: 0.70 (20 °C)
Flash Pt: 14 °C
Storage Temperature: Ambient
MDL Number: MFCD00009556
CAS Number: 111-65-9
EINECS: 203-892-1
UN: 1262
ADR: 3,II
Merck Index: 13,06782
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=3d1aad7bdf5c4ca241411dda5ad0d0cdd320c89f&vid=02732d3d1626619b13bbb3effbbd1ca92ee7a226&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.5% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR