1,2-Epoxycyclohexane ≥98.0%
Supplier: TCI America
|
Danger
|
Synonyms:
Epoxycyclohexane, Cyclohexene oxide
O0071-500ML
O0071-25ML
O0071-100ML
TCO0071-500MLEA
82
USD
TCO0071-500ML
TCO0071-25ML
TCO0071-100ML
1,2-Epoxycyclohexane ≥98.0%
1,2-Epoxycyclohexane
CAS Number: 286-20-4
MDL Number: MFCD00005162
Molecular Formula: C6H10O
Molecular Weight: 98.15
Purity/Analysis Method:
Other Product Names / Synonyms: Cyclohexene Oxide, 7-Oxabicyclo[4.1.0]heptane
Formula:
C₆H₁₀O MW: 98.14 g/mol Boiling Pt: 129 Melting Pt: -27 Density: 0.97 (20 °C) Flash Pt: 27 °C Storage Temperature: Ambient |
MDL Number:
MFCD00005162 CAS Number: 286-20-4 EINECS: 206-007-7 UN: 2920 ADR: 8 II |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...