Oleyl chloride ≥70.0%
Supplier: TCI America
O0060-25ML
TCO0060-025MLEA
75
USD
TCO0060-025ML
TCO0060-500ML
Oleyl chloride ≥70.0%
Oleyl chloride
CAS Number: 16507-61-2
MDL Number: MFCD00058939
Molecular Formula: C18H35Cl
Molecular Weight: 286.93
Purity/Analysis Method:
Formula:
C₁₈H₃₅Cl Density: 0.87 (20 °C) |
MDL Number:
MFCD00058939 CAS Number: 16507-61-2 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...