Ethyl oleate ≥70.0%
Supplier: TCI America
Synonyms:
Elaidic acid ethyl ester, (Z)-9-Octadecenoic acid ethyl ester, Oleic acid ethyl ester
O0054-25ML
O0054-500ML
TCO0054-025MLEA
27.48
USD
TCO0054-025ML
TCO0054-500ML
Ethyl oleate ≥70.0%
Ethyl oleate
CAS Number: 111-62-6
MDL Number: MFCD00009579
Molecular Formula: C20H38O2
Molecular Weight: 310.52
Purity/Analysis Method:
Product Categories: Materials Chemistry, Resin Additives, Bioscience, Lipids, Higher Fatty Acids & Higher Alcohols
Other Product Names / Synonyms: Oleic Acid Ethyl Ester
Formula:
C₂₀H₃₈O₂ Boiling Pt: 209 °C Melting Pt: –32 °C Density: 0.87 (20 °C) Flash Pt: 181 °C Storage Temperature: Ambient |
MDL Number:
MFCD00009579 CAS Number: 111-62-6 EINECS: 203-889-5 Merck Index: 12,06965 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...