Necrostatin-1 ≥98.0% (by HPLC)

Supplier: TCI America

Synonyms: 5-((1H-Indol-3-yl)methyl)-3-methyl-2-thioxoimidazolidin-4-one, Nec-1

N1174-100MG
TCN1174-100MGEA 314.15 USD
TCN1174-100MG
Necrostatin-1 ≥98.0% (by HPLC)
Necrostatin-1
Formula: C₁₃H₁₃N₃OS
MW: 259.327 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD00056916
CAS Number: 4311-88-0

Order Now

Specification Test Results

Appearance Light orange to Yellow to Green powder to crystal
Purity (HPLC) min. 98.0 area%
Melting point 160.0 to 164.0 deg-C
Elemental analysis (Nitrogen) 15.40 to 17.00 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR